6-ROXtra™ acid fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
386
AATB386EA
426
EUR
AATB386
6-ROXtra™ acid fluoreszierende Probe
6-ROXtra™ acid
Although ROX dyes have strong fluorescence they are notoriously unstable.
- Improved water solubility
- Easily purified
- Strong fluorescence
Molecular Weight:
1034,34 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...