5(6)-ROX N-succinimidyl ester Gemisch aus Isomeren ≥90%
Lieferant: AAT BIOQUEST
Synonyme:
5(6)-ROX, SE, Carboxy-X-rhodamine succinimidyl ester, 5 and 6 isomers, 5(6)-Carboxy-X-rhodamine N-succinimidyl ester
390
AATB390EA
307
EUR
AATB390
5(6)-ROX N-succinimidyl ester Gemisch aus Isomeren ≥90%
5(6)-ROX N-succinimidyl ester
5(6)-ROX, SE is the succinimidyl esters of 5(6)-ROX.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.
Formel:
C₃₇H₃₃N₃O₇ Molecular Weight: 631,67 g/mol |
CAS-Nummer:
114616-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...