SDA azide
Lieferant: AAT Bioquest
39004
AATB39004EA
133
EUR
AATB39004
SDA azide
SDA azide
SDA azide contains both diazirine photoaffinity moiety and azide functional group.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only