SDA azide
Lieferant: AAT Bioquest
39004
AATB39004EA 133 EUR
AATB39004
SDA azide
SDA azide
SDA azide contains both diazirine photoaffinity moiety and azide functional group.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only