SDA alkyne
Lieferant: AAT BIOQUEST
39005
AATB39005EA
133
EUR
AATB39005
SDA alkyne
SDA alkyne
SDA alkyne contains both diazirine photoaffinity moiety and alkyne functional group.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only