SDA maleimide
Lieferant: AAT Bioquest
39006
AATB39006EA 133 EUR
AATB39006
SDA maleimide
SDA maleimide
SDA maleimide is an excellent heterobifunctional crosslinker that effectively combines two established chemical modalities, thiol-selective maleimide chemistry and diazirine-based photoaffinity labeling.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only