SDA maleimide
Lieferant: AAT Bioquest
SDA maleimide is an excellent heterobifunctional crosslinker that effectively combines two established chemical modalities, thiol-selective maleimide chemistry and diazirine-based photoaffinity labeling.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only