Trifluoromethyl phenyl diazirine NHS ester
Lieferant: AAT BIOQUEST
39007
AATB39007EA 271 EUR
AATB39007
Trifluoromethyl phenyl diazirine NHS ester
Trifluoromethyl phenyl diazirine NHS ester
Trifluoromethylphenyldiazirines are one type of the most effective diazirines with generally high photoreaction reaction yield by UV irradiation (~350 nm).
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only