Trifluoromethyl phenyl diazirine maleimide
Lieferant: AAT BIOQUEST
39008
AATB39008EA
271
EUR
AATB39008
Trifluoromethyl phenyl diazirine maleimide
Trifluoromethyl phenyl diazirine maleimide
Trifluoromethyl phenyl diazirine maleimide is an excellent building block to develop photoactive probes by reacting with a thiol-containing biomolecule.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only