Trifluoromethyl phenyl diazirine amine
Lieferant: AAT BIOQUEST
39009
AATB39009EA
133
EUR
AATB39009
Trifluoromethyl phenyl diazirine amine
Trifluoromethyl phenyl diazirine amine
Trifluoromethylphenyldiazirines are one type of the most effective diazirines with generally high photoreaction reaction yield by UV irradiation (~350 nm).
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only