4-Azido-2,3,5,6-tetrafluorobenzoic acid succinimidyl ester
Lieferant: AAT BIOQUEST
39010
AATB39010EA
133
EUR
AATB39010
4-Azido-2,3,5,6-tetrafluorobenzoic acid succinimidyl ester
4-Azido-2,3,5,6-tetrafluorobenzoic acid succinimidyl ester
Phenyl azides are a well-known class of photoaffinity probes that require photoactivation at short wavelengths (∼260 nm).
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only