Psoralen MOP succinimidyl ester
Lieferant: AAT BIOQUEST
39055
AATB39055EA 266 EUR
AATB39055
Psoralen MOP succinimidyl ester
Psoralen MOP succinimidyl ester
Psoralens and their derivatives (such as 8-MOP and 4,5’8-TMP) are well known to have unique crosslinking features to DNA.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only