Psoralen MOP maleimide
Lieferant: AAT BIOQUEST
39056
AATB39056EA 266 EUR
AATB39056
Psoralen MOP maleimide
Psoralen MOP maleimide
Psoralens and their derivatives (such as 8-MOP and 4,5’8-TMP) are well known to have unique crosslinking features to DNA.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only