Psoralen MOP alkyne
Lieferant: AAT BIOQUEST
39058
AATB39058EA 266 EUR
AATB39058
Psoralen MOP alkyne
Psoralen MOP alkyne
Psoralen MOP Alkyne is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Azide–Alkyne Cycloaddition (CuAAC).
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only