Psoralen TMP succinimidyl ester
Lieferant: AAT BIOQUEST
39060
AATB39060EA
266
EUR
AATB39060
Psoralen TMP succinimidyl ester
Psoralen TMP succinimidyl ester
Psoralens and their derivatives (such as 8-TMP and 4,5’,8-TMP) are well known to have unique crosslinking features to DNA.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only