Psoralen TMP alkyne
Lieferant: AAT BIOQUEST
39063
AATB39063EA 266 EUR
AATB39063
Psoralen TMP alkyne
Psoralen TMP alkyne
Psoralen TMP Alkyne is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Azide–Alkyne Cycloaddition (CuAAC).
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Environmentally Preferable