Sunnyvale Red™ SE *Superior 6-ROX Replacement*
Lieferant: AAT BIOQUEST
394
AATB394EA
307
EUR
AATB394
Sunnyvale Red™ SE *Superior 6-ROX Replacement*
Tests
qPCR-Assays
Although ROX dyes have strong fluorescence they are notoriously unstable.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.