6-ROX glycine-Lösung 25 µM Referenzstandard, für PCR
Lieferant: AAT BIOQUEST
395
AATB395EA
307
EUR
AATB395
6-ROX glycine-Lösung 25 µM Referenzstandard, für PCR
6-ROX glycine
6-ROX is predominately used as a reference dye for performing PCR detections.
- Multiple Bioconjugation Applications
- 95% Purity
Bitte beachten For research use only.