6-ROX glycine-Lösung 25 µM Referenzstandard, für PCR
Lieferant: AAT BIOQUEST
395
AATB395EA 307 EUR
AATB395
6-ROX glycine-Lösung 25 µM Referenzstandard, für PCR
6-ROX glycine
6-ROX is predominately used as a reference dye for performing PCR detections.
  • Multiple Bioconjugation Applications
  • 95% Purity
Bitte beachten For research use only.