6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*
Lieferant: AAT BIOQUEST
398
AATB398EA
460
EUR
AATB398
6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*
Tests
qPCR-Assays
6-ROX is predominately used as a reference dye for performing PCR detections.
- Multiple bioconjugation applications
- 95% purity
Bitte beachten For research use only.