3-Cyanoumbelliferone ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
3-Cyano-7-hydroxycoumarin, 7-Hydroxy-2-oxo-2H-chromene-3-carbonitrile
40
AATB40EA
120
EUR
AATB40
3-Cyanoumbelliferone ≥95%
3-Cyanoumbelliferone
3-Cyano-7-hydroxycoumarin is used as a calibration standard for 3-cyano-7-hydroxycoumarin-based enzyme substrates.
- pH-dependent fluorescence
Formel:
C₁₀H₅NO₃ Molecular Weight: 187,15 g/mol |
MDL:
MFCD00037480 CAS-Nummer: 19088-73-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...