Amplite® Ethanol Quantitation Kit
Lieferant: AAT BIOQUEST
40001
AATB40001EA
307
EUR
AATB40001
Amplite® Ethanol Quantitation Kit
Tests
Enzymtests
The ability to rapidly perform quantitative measurements of ethanol is highly desirable in life science research, clinical evaluations, and food and drug industries.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.