6-Amino-1-hexanol ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
6-Aminohexan-1-ol
4001
AATB4001EA
152
EUR
AATB4001
6-Amino-1-hexanol ≥95%
6-Amino-1-hexanol
6-Aminohexanol is a precursor that is widely used for preparing biotin and dye phosphoramidites.
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol |
MDL:
MFCD00008241 CAS-Nummer: 4048-33-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...