6-Amino-1-hexanol zur Proteinmarkierung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Aminohexan-1-ol

4002
AATB4002EA 537 EUR
AATB4002
6-Amino-1-hexanol zur Proteinmarkierung
6-Amino-1-hexanol

6-Aminohexanol is a precursor that is widely used for preparing biotin and dye phosphoramidites.


  • High quality
  • Phosphoramidite precursor

Formel: C₆H₁₅NO
Molecular Weight: 117,19 g/mol
MDL: MFCD00008241
CAS-Nummer: 4048-33-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b528d876759447f9d4caa3eead4e975ac95b33&vid=ccefff95aa5bcaec5bf881cc16fd08819ca47e2d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR