3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride zur Oligonukleotidmarkierung

Lieferant: AAT BIOQUEST
Achtung

4021
AATB4021EA 460 EUR
AATB4021
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride zur Oligonukleotidmarkierung
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride

3,6-Dichlorotrimellitic anhydride is the key precursor that is used for preparing a variety of dichlorinated fluoresceins and rhodamines such as TET and HEX.


  • Bioconjugate building block
  • Multiple applications

Formel: C₉H₂Cl₂O₅
MDL: MFCD28962732
CAS-Nummer: 81742-10-1
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b61bb634ad79607ca8f64381ef5ac0baa637f70f&vid=55a338e2aec421c71ff2b26c02c402af1f8fb68a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR