4-Fluororesorcinol
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
1-Fluoro-2,4-dihydroxybenzene
4022
AATB4022EA
152
EUR
AATB4022
4-Fluororesorcinol
4-Fluororesorcinol
4-Fluororesorcinol is a precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs.
Formel:
C₆H₅FO₂ Molecular Weight: 128,10 g/mol |
MDL:
MFCD03789074 CAS-Nummer: 103068-41-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...