2,4-Difluorresorcin ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2,4-Difluorobenzene-1,3-diol

4023
AATB4023EA 230 EUR
AATB4023
2,4-Difluorresorcin ≥90%
2,4-Difluorresorcin

2,4-Difluororesorcinol is a precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs.

Formel: C₆H₄F₂O₂
CAS-Nummer: 195136-71-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR