4-Methylumbelliferon Fluoreszenz-Referenzstandard

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 7-Hydroxy-4-methylcumarin, Hymecromon

41
AATB41EA 120 EUR
AATB41
4-Methylumbelliferon Fluoreszenz-Referenzstandard
4-Methylumbelliferon

This compound is used as a calibration standard for 7-hydroxy-4-methylcoumarin-based enzyme substrates.


  • pH-dependent fluorescence

Formel: C₁₀H₈O₃
Molecular Weight: 176,17 g/mol
MDL: MFCD00006866
CAS-Nummer: 90-33-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64e91c9b42debf9a737d6f1483d1daba47020b70&vid=77bd39318f1a0a01ff57eb9b8df0b8c55e50a109&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR