5(6)-TAMRA Gemisch aus Isomeren ≥85%
Lieferant: AAT BIOQUEST
Synonyme:
TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt
412
AATB412EA
307
EUR
AATB412
5(6)-TAMRA Gemisch aus Isomeren ≥85%
5(6)-TAMRA
Tetramethylrhodamine-5-(and-6)-maleimide is an excellent reagent for preparing orange fluorescent bioconjugates through thiol modifications of peptides, nucleotides, nucleic acids and proteins.
- Multiple Bioconjugation Applications
- 85% Purity
Bitte beachten For research use only.
Formel:
C₂₅H₂₂N₂O₅ Molecular Weight: 430,45 g/mol |
MDL:
MFCD16661194 CAS-Nummer: 98181-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...