Tetramethylrhodamine-5-isothiocyanate (5-TRITC) ≥90%

Lieferant: AAT Bioquest
Achtung

415 416
AATB415EA 307 EUR
AATB415 AATB416
Tetramethylrhodamine-5-isothiocyanate (5-TRITC) ≥90%
Tetramethylrhodamine-5-isothiocyanate (5-TRITC)

5-TRITC (also called G isomer) is a purified single isomer of the 5(6)-TRITC mixed isomers.

Formel: C₂₅H₂₁N₃O₃S
MDL: MFCD07438035
CAS-Nummer: 80724-19-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65d8fae5f754a2b8f418940d39adcc909d7260f5&vid=e230e37e95f58ed5a5536a29114ca59247ba45e3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR