Tetramethylrhodamine-6-maleimide ≥90%
Lieferant: AAT Bioquest
|
Achtung
|
419
AATB419EA
307
EUR
AATB419
Tetramethylrhodamine-6-maleimide ≥90%
Tetramethylrhodamine-6-maleimide
Tetramethylrhodamine-6-maleimide is the other purified single isomer of tetramethylrhodamine-5 (and 6)-maleimide mixed isomers.
Formel:
C₂₈H₂₃N₃O₅ |
CAS-Nummer:
174568-68-4 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...