5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide] ≥90%
Lieferant: AAT Bioquest
Achtung
421
AATB421EA 307 EUR
AATB421
5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide] ≥90%
5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide]
Tetramethylrhodamine-5-maleimide is the purified single isomer of tetramethylrhodamine-5 (and 6)-maleimide mixed isomers.
CAS-Nummer: 154480-30-5
ADR: 5,II