6-TAMRA C6 maleimide ≥90%
Lieferant: AAT Bioquest
425
AATB425EA
307
EUR
AATB425
6-TAMRA C6 maleimide ≥90%
6-TAMRA C6 maleimide
6-TAMRA C6 maleimide is an excellent replacement for tetramethylrhodamine-5-maleimide.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.