6-ROX C2 maleimide ≥90%
Lieferant: AAT BIOQUEST
426
AATB426EA 152 EUR
AATB426
6-ROX C2 maleimide ≥90%
6-ROX C2 maleimide
6-ROX C2 maleimide is an excellent thiol-reactive ROX probe that can be used to develop new ROX bioconjugates.
  • Multiple Bioconjugation Applications
  • 90% Purity
Bitte beachten For research use only.