FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimised for Tyramide and Styramide Imaging*
Lieferant: AAT BIOQUEST
44890
AATB44890EA
153
EUR
AATB44890
FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimised for Tyramide and Styramide Imaging*
Färbemittel
Mikroskopie- und Färbereagenzien
FluoroQuest™ TSA/PSA Antifade mounting medium is the only mounting medium that is optimised for tyramide (TSA) and styramide (PSA)-based fluorescence imaging applications.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.