iFluor® 660 styramide
Lieferant: AAT BIOQUEST
44906
AATB44906EA 429 EUR
AATB44906
iFluor® 660 styramide
iFluor® 660 styramide
iFluor® 660 dyes have fluorescence excitation and emission maxima of ~660 nm and ~680 nm respectively.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Bitte beachten For Research Use Only