iFluor® 450, Reactive Form_Styramide *Superior Replacement for Opal Polaris 480*
Lieferant: AAT BIOQUEST
45010
AATB45010EA 426 EUR
AATB45010
iFluor® 450, Reactive Form_Styramide *Superior Replacement for Opal Polaris 480*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
  • 10 to 50× higher fluorescence
  • improved precision and sensitivity
  • more efficient use of primary antibody
  • enhanced water solubility
  • more photostable
Bitte beachten For research use only.