iFluor® 450, Reactive Form_Styramide *Superior Replacement for Opal Polaris 480*
Lieferant: AAT BIOQUEST
45010
AATB45010EA
426
EUR
AATB45010
iFluor® 450, Reactive Form_Styramide *Superior Replacement for Opal Polaris 480*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
- 10 to 50× higher fluorescence
- improved precision and sensitivity
- more efficient use of primary antibody
- enhanced water solubility
- more photostable
Bitte beachten For research use only.