iFluor® 514 styramide [Superior Replacement for Opal 540]
Lieferant: AAT BIOQUEST
45022
AATB45022EA 429 EUR
AATB45022
iFluor® 514 styramide [Superior Replacement for Opal 540]
iFluor® 514 styramide
iFluor® 514 Styramide is optimized to be a superior replacement for Opal Polaris 540 or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Bitte beachten For Research Use Only