iFluor® 546, Styramid "Superior Replacement for Alexa Fluor 546 tyramide"
Lieferant: AAT BIOQUEST
45025
AATB45025EA
460
EUR
AATB45025
iFluor® 546, Styramid "Superior Replacement for Alexa Fluor 546 tyramide"
iFluor®
Power Styramide™ signal amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
- 10 to 50× higher fluorescence
- Improved precision and sensitivity
- More efficient use of primary antibody
- Enhanced water solubility
- More photostable
Bitte beachten For research use only.