iFluor® 594, Styramid *Superior Replacement for Alexa Fluor 594 tyramide*
Lieferant: AAT BIOQUEST
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents.
- 10 to 50× higher fluorescence
- Improved precision and sensitivity
- More efficient use of primary antibody
- Enhanced water solubility
- More photostable
Bitte beachten For research use only.