BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS
4507
AATB4507EA
152
EUR
AATB4507
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a non-cleavable and membrane permeable crosslinker.
Formel:
C₁₁H₁₀N₂O₆ Molecular Weight: 266,21 g/mol |
MDL:
MFCD00043141 CAS-Nummer: 55750-62-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...