mFluor™ Blue 580 styramide
Lieferant: AAT BIOQUEST
45071
AATB45071EA 420 EUR
AATB45071
mFluor™ Blue 580 styramide
mFluor™ Blue 580 styramide
mFluor™ Blue 580 Styramide uses the bright red-orange fluorescent dye mFluor™ Blue 580 (Ex/Em = 485/580 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only