mFluor™ Blue 630 styramide
Lieferant: AAT Bioquest
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only