mFluor™ Blue 630 styramide
Lieferant: AAT Bioquest
45073
AATB45073EA 420 EUR
AATB45073
mFluor™ Blue 630 styramide
mFluor™ Blue 630 styramide
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only