mFluor™ Blue 660 styramide
Lieferant: AAT BIOQUEST
45074
AATB45074EA
420
EUR
AATB45074
mFluor™ Blue 660 styramide
mFluor™ Blue 660 styramide
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only