mFluor™ Green 620 styramide
Lieferant: AAT Bioquest
mFluor™ Green 620 Styramide uses the bright red fluorescent dye mFluor™ Green 620 (Ex/Em = 525/623 nm) to label targets in situ.
- Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only