mFluor™ Green 620 styramide
Lieferant: AAT Bioquest
45075
AATB45075EA 420 EUR
AATB45075
mFluor™ Green 620 styramide
mFluor™ Green 620 styramide
mFluor™ Green 620 Styramide uses the bright red fluorescent dye mFluor™ Green 620 (Ex/Em = 525/623 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only