mFluor™ Red 780 styramide
Lieferant: AAT BIOQUEST
45076
AATB45076EA 420 EUR
AATB45076
mFluor™ Red 780 styramide
mFluor™ Red 780 styramide
mFluor™ Red 780 Styramide uses the bright red fluorescent dye mFluor™ Red 780 (Ex/Em = 629/767 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only