mFluor™ Violet 540 styramide
Lieferant: AAT BIOQUEST
45077
AATB45077EA 420 EUR
AATB45077
mFluor™ Violet 540 styramide
mFluor™ Violet 540 styramide
mFluor™ Violet 540 Styramide uses the bright green fluorescent dye mFluor™ Violet 540 (Ex/Em = 402/535 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only