ReadiUse™ Tyramide (TSA)/Styramide (PSA) Optimized Reaction Buffer (1000 Reactions)
Lieferant: AAT BIOQUEST
45090
AATB45090EA
143
EUR
AATB45090
ReadiUse™ Tyramide (TSA)/Styramide (PSA) Optimized Reaction Buffer (1000 Reactions)
Puffer
Assay-Puffer
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
- Improved fluorescent signal
- Suitable for either TSA or PSA reactions
- Ready to use
Bitte beachten For research use only.