iFluor® 488, Tyramid
Lieferant: AAT BIOQUEST
45100
AATB45100EA 460 EUR
AATB45100
iFluor® 488, Tyramid
iFluor®
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
Bitte beachten For research use only.