iFluor® 546, Tyramid
Lieferant: AAT BIOQUEST
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
- Increased fluorescence signal
- Suitable for low-abundance targets
- Compatible with all major fluorescence platforms
Bitte beachten For research use only.