iFluor® 594, Tyramid
Lieferant: AAT BIOQUEST
45107
AATB45107EA 439 EUR
AATB45107
iFluor® 594, Tyramid
iFluor®
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
Bitte beachten For research use only.