DSG (Disuccinimidyl glutarate) ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
DSG
4520
AATB4520EA
120
EUR
AATB4520
DSG (Disuccinimidyl glutarate) ≥90%
DSG (Disuccinimidyl glutarate)
DSG is disuccinimidyl glutarate, a simple homobifunctional crosslinker based on the amine-reactive N-hydroxysuccinimide (NHS) ester group at both ends of a 5-atom (7.7 angstrom) spacer arm.
Formel:
C₁₃H₁₄N₂O₈ Molecular Weight: 326.26 g/mol |
MDL:
MFCD00153597 CAS-Nummer: 79642-50-5 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...