m-PEG24-Mal zur biomolekularen Markierung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Methyl-PEG24-Maleimide, m-PEG24-Maleimide

4525
AATB4525EA 152 EUR
AATB4525
m-PEG24-Mal zur biomolekularen Markierung
m-PEG24-Mal

MM(PEG)24 is a methyl-terminated, polyethylene glycol compound (24 PEG units) activated with a maleimide group for covalent pegylation of sulfhydryls on biomolecules or assay surfaces.


  • Improved crosslinking efficiency
  • High water solubility

Formel: C₅₆H₁₀₆N₂O₂₇
Molecular Weight: 1239,45 g/mol
MDL: MFCD11041152
CAS-Nummer: 88504-24-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a67947e77909c3e417badc56cd989beebbddf1&vid=b2f4a50e696649eb46ef4b12e955a7e241cecc8e&prid=web&clid=SAPDAM
Neuer Artikel bei VWR!

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR